Assay Details
| Gene Symbol: 4930532J02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AA839396.1 | - | 1 | 307 | 77 | |
| AK015945.1 | - | 1 | 253 | 77 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
regulatory factor X, 5 (influences HLA class II expression) |
Rfx5 | Anti-sense | Chr. 3 - 94951957 - 94961561 on NCBI Build GRCm38 | ||
| Downstream | RIKEN cDNA B230398E01 gene | B230398E01Rik | 6720476P12Rik | 5.7kb | Sense | Chr. 3 - 94959674 - 94986687 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930532J02Rik
Genomic Map