Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200013836.mm.1 |
NR_038349.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013836.mm.1 |
NR_038349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013836.mm.1 |
TC0200001799.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200013844.mm.1 |
NR_038349.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013844.mm.1 |
NR_038349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013844.mm.1 |
NR_038350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013844.mm.1 |
NR_038350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013844.mm.1 |
TC0200001799.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200013846.mm.1 |
NR_038349.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013846.mm.1 |
NR_038349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013846.mm.1 |
NR_038350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013846.mm.1 |
NR_038350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013846.mm.1 |
TC0200001799.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200013842.mm.1 |
NR_038349.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013842.mm.1 |
NR_038349.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200013842.mm.1 |
TC0200001799.mm.1 |
MTA TC |