Assay Details
Gene Symbol: 4930535I16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK015982.1 | - | 1 | 650 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | c-myc binding protein | Mycbp |
5730488M09Rik, AW552132, AW742590, Amy-1 |
-4.7kb | Anti-sense | Chr. 4 - 123905013 - 123912250 on NCBI Build GRCm38 |
Downstream | predicted gene 12916 | Gm12916 | OTTMUSG00000009178 | 375.1kb | Sense | Chr. 4 - 124293102 - 124299923 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
