Assay Details
| Gene Symbol: 4930554G22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016118.1 | - | 1 | 252 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | colony stimulating factor 1 (macrophage) | Csf1 |
C87615, Csfm, MCSF, op |
-114.3kb | Anti-sense | Chr. 3 - 107741048 - 107760469 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4933431E20 gene | 4933431E20Rik | 13.4kb | Anti-sense | Chr. 3 - 107888850 - 107896213 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930554G22Rik
Genomic Map