Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200006994.mm.1 |
NR_040633.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006994.mm.1 |
NR_040633.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006994.mm.1 |
TC0200000867.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006993.mm.1 |
NR_040633.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006993.mm.1 |
NR_040633.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006993.mm.1 |
TC0200000867.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200006995.mm.1 |
NR_040633.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006995.mm.1 |
NR_040633.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200006995.mm.1 |
TC0200000867.mm.1 |
MTA TC |