Assay Details
| Gene Symbol: 4930566K23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK016238.1 | - | 1 | 546 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 16731 |
Gm16731 | -27.3kb | Sense | Chr. 4 - 46539092 - 46539883 on NCBI Build GRCm38 | |
| Downstream |
TBC1 domain family, member 2 |
Tbc1d2 |
A630005A06Rik, BC022630, Gm1037, PARIS-1, PARIS1 |
36.5kb | Anti-sense | Chr. 4 - 46604390 - 46650877 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

4930566K23Rik
Genomic Map