Assay Details
Gene Symbol: 4930588G17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK016366.1 | - | 1 - 2 | 440 | 102 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | PDZ domain containing 9 | Pdzd9 | 4930408O21Rik | Anti-sense | Chr. 7 - 120659104 - 120677562 on NCBI Build GRCm38 | |
Downstream | cDNA sequence BC030336 | BC030336 | 6.1kb | Sense | Chr. 7 - 120677620 - 120734854 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
