Assay Details
Gene Symbol: 4930500F04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK016439.1 | - | 1 - 2 | 168 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 21221 |
Gm21221 | -40.5kb | Anti-sense | Chr. 5 - 149801990 - 149811751 on NCBI Build GRCm38 | |
Downstream | relaxin/insulin-like family peptide receptor 2 | Rxfp2 |
Gpr106, Great, Lgr8 |
163.4kb | Sense | Chr. 5 - 150017781 - 150082550 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
