Assay Details
| Gene Symbol: 4933408J17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_045810.1 | 2 - 3 | 832 | 86 | ||
| GenBank mRNA | AK016739.1 | - | 2 - 3 | 835 | 86 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | small nuclear ribonucleoprotein polypeptide F | Snrpf |
2010013O18Rik, SMF, sm-F, snRNP-F |
-6.2kb | Anti-sense | Chr. 10 - 93583029 - 93589658 on NCBI Build GRCm38 |
| Downstream | netrin 4 | Ntn4 | 41.5kb | Sense | Chr. 10 - 93641049 - 93745972 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4933408J17Rik
Genomic Map