Assay Details
| Gene Symbol: 5230400M06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017228.1 | - | 1 | 1752 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 503 | Zfp503 |
AI181838, B830002A16Rik, Nolz-1, Nolz1, Znf503 |
-411.5kb | Anti-sense | Chr. 14 - 21983962 - 21989601 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4930405A10 gene | 4930405A10Rik | 4930456G20Rik | 104.2kb | Anti-sense | Chr. 14 - 22507174 - 22542798 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5230400M06Rik
Genomic Map