Assay Details
Gene Symbol: 5430402P08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK017263.1 | - | 1 - 2 | 209 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 1700012D14 gene | 1700012D14Rik | -243.6kb | Anti-sense | Chr. 7 - 111117673 - 111122675 on NCBI Build GRCm38 | |
Downstream | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 18 | Galnt18 |
2900011G21Rik, BC024988, Galntl4 |
76.0kb | Anti-sense | Chr. 7 - 111471656 - 111780067 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2383083 | AK017263.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK011431.2 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK044933.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK088075.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK151336.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK151658.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK151661.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK151869.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK152026.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK152453.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK152455.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK152911.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK153294.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK153371.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK159338.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK159807.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK162426.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK163244.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK166605.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK167828.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK168698.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK168931.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK169042.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | AK169226.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | BC002052.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | BC021768.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | BC037730.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | NM_008211.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931224 | X13605.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 786055 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 9666 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 857963 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 79385 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939012 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 519709 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179781 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 22903 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10764 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119883 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119883 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06322686 | - | - |
Whole Mouse Genome G4122A | A_52_P826044 | AK017263.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107091 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119883 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418472 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418338 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418481 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418466 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418326 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06552242 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418482 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418382 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418378 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418381 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418324 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418342 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418392 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418327 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418383 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418458 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06449227 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418390 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418340 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418463 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06552235 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418388 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418473 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418389 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418462 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05891186 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418328 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418467 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418377 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418337 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418336 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418323 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418376 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418333 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418475 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06647065 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418322 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418331 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418471 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418483 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418379 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25418478 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1432248_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1432248_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1432248_at | - | - |
Mouse Expression Set 430 | 1432248_PM_at | - | - |
Mouse Expression Set 430 | 1432248_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432248_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1432248_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 617251 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 106840136 | - | - |
