Assay Details
| Gene Symbol: Sh3bgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017399.1 | - | 1 | 952 | 55 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Leber congenital amaurosis 5-like | Lca5l |
4921526F01Rik, Lcca5l |
-6.7kb | Anti-sense | Chr. 16 - 96158405 - 96193993 on NCBI Build GRCm38 |
| Downstream |
UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 5 |
B3galt5 |
1190002B21Rik, AU045265, b3Galt-V |
34.0kb | Sense | Chr. 16 - 96235801 - 96319859 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sh3bgr
Genomic Map