Assay Details
| Gene Symbol: 5530402G07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017444.1 | - | 1 | 654 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 5124a | Mir5124a |
Mir5124, mmu-mir-5124, mmu-mir-5124a, mu-mir-5124 |
-83.0kb | Anti-sense | Chr. 13 - 40865791 - 40865862 on NCBI Build GRCm38 |
| Downstream | PAK1 interacting protein 1 | Pak1ip1 |
5830431I15Rik, 5930415H02Rik, AA419825, AI314040, AW556169, Gdpd1, PIP1 |
51.4kb | Anti-sense | Chr. 13 - 41001010 - 41013033 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5530402G07Rik
Genomic Map