Assay Details
| Gene Symbol: 5730409N24Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI606154.1 | - | 1 | 92 | ||
| AK017539.1 | - | 1 | 1556 | 92 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sorbin and SH3 domain containing 1 | Sorbs1 |
2310065E01Rik, 9530001P15Rik, CAP, SH3P12, Sh3d5, mKIAA1296 |
Sense | Chr. 19 - 40292040 - 40513876 on NCBI Build GRCm38 | |
| Downstream |
aldehyde dehydrogenase 18 family, member A1 |
Aldh18a1 |
2810433K04Rik, AI429789, Pycs |
75.8kb | Sense | Chr. 19 - 40550257 - 40588463 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5730409N24Rik
Genomic Map