Assay Details
Gene Symbol: D10Bwg1070e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AA671314.1 | - | 1 | 262 | 84 | |
AK017596.1 | - | 1 | 351 | 84 | ||
AK158707.1 | - | 1 | 1200 | 84 | ||
BE981800.1 | - | 1 | 572 | 84 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | heterogeneous nuclear ribonucleoprotein H3 | Hnrnph3 |
AA693301, AI666703, Hnrph3 |
Sense | Chr. 10 - 63014650 - 63023889 on NCBI Build GRCm38 | |
Downstream | phenazine biosynthesis-like protein domain containing 2 | Pbld2 |
3110049J23Rik, Mawbp |
1.1kb | Anti-sense | Chr. 10 - 63024049 - 63058813 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
