Assay Details
| Gene Symbol: 5730433N10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017612.1 | - | 1 | 1257 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
serine (or cysteine) peptidase inhibitor, clade E, member 2 |
Serpine2 |
B230326M24Rik, PAI-1, PI-7, PI7, PN-1, Spi4 |
Sense | Chr. 1 - 79794319 - 79861175 on NCBI Build GRCm38 | |
| Downstream |
family with sequence similarity 124, member B |
Fam124b | A830043J08Rik | 346.6kb | Sense | Chr. 1 - 80186969 - 80218504 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5730433N10Rik
Genomic Map