Assay Details
Gene Symbol: Gpatch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK017966.1 | - | 1 | 144 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | spermatogenesis associated 17 | Spata17 |
1700065F16Rik, 4930504I07Rik, 4930513F16Rik, MSRG-11 |
-21.0kb | Anti-sense | Chr. 1 - 187009614 - 187215462 on NCBI Build GRCm38 |
Downstream |
predicted gene, 37896 |
Gm37896 | A930038G18 | 210.0kb | Sense | Chr. 1 - 187447697 - 187462781 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
