Assay Details
| Gene Symbol: 5830435N06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017970.1 | - | 1 | 1146 | 58 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microfibrillar-associated protein 3 | Mfap3 |
2610509F16Rik, 2700079M14Rik, AW536327 |
-131.9kb | Sense | Chr. 11 - 57518652 - 57533817 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 2010001A14 gene | 2010001A14Rik | 128.3kb | Anti-sense | Chr. 11 - 57795495 - 57801534 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5830435N06Rik
Genomic Map