Assay Details
| Gene Symbol: 5830485P09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018061.1 | - | 1 | 1455 | 93 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | thymopoietin | Tmpo |
5630400D24Rik, AI195756, AI606875, AW214352, AW547477, LAP2, TP |
Sense | Chr. 10 - 91147571 - 91181719 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 31592 |
Gm31592 | 736.9kb | Anti-sense | Chr. 10 - 91888586 - 91953620 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

5830485P09Rik
Genomic Map