Assay Details
| Gene Symbol: 6330575P09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018252.1 | - | 1 | 1053 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | serine/threonine kinase 4 | Stk4 |
AI447339, AU020804, Kas-2, Mst1, Ysk3 |
Sense | Chr. 2 - 164074109 - 164155524 on NCBI Build GRCm38 | |
| Downstream |
K+ voltage-gated channel, subfamily S, 1 |
Kcns1 | Kv9.1 | 71.3kb | Anti-sense | Chr. 2 - 164163619 - 164171113 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6330575P09Rik
Genomic Map