Assay Details
| Gene Symbol: 6430514K02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018281.1 | - | 1 | 953 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | teneurin transmembrane protein 3 | Tenm3 |
2610100B16Rik, Odz1, Odz3, Ten-m3, mKIAA1455 |
-803.3kb | Sense | Chr. 8 - 48225665 - 48935162 on NCBI Build GRCm38 |
| Downstream | predicted gene 2516 | Gm2516 | 628.2kb | Anti-sense | Chr. 8 - 50368112 - 50416089 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 872783 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06508218 | - | - |
| Whole Mouse Genome G4122A | A_51_P217759 | AK018281.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06508218 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856201 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856171 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07178569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856264 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856173 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856202 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856190 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856200 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856144 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856186 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856175 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856188 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856137 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856184 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856163 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856181 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856177 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856258 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856193 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07273400 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856159 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856199 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856261 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856255 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856198 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856127 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856158 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856170 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856141 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856196 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856174 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856187 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856179 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856126 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856165 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856155 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856160 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856139 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856265 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07102110 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856132 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856183 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856129 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856125 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856259 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856156 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06737894 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856168 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856169 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856257 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856172 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856263 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856134 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25856197 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT MG-430 PM Array Plate | 1433091_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1433091_at | - | - |
| Mouse Genome 430 2.0 Array | 1433091_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1433090_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1433090_at | - | - |
| Mouse Expression Set 430 | 1433091_PM_at | - | - |
| Mouse Expression Set 430 | 1433091_at | - | - |
| Mouse Expression Set 430 | 1433090_at | - | - |
| Mouse Expression Set 430 | 1433090_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433091_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1433091_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 102320167 | - | - |

6430514K02Rik
Genomic Map