Assay Details
| Gene Symbol: 6430711C07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018304.1 | - | 1 | 1551 | 83 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 20756 |
Gm20756 | -1.9mb | Anti-sense | Chr. 6 - 25931426 - 25977227 on NCBI Build GRCm38 | |
| Downstream | microRNA 592 | Mir592 |
Mirn592, mir-592, mmu-mir-592 |
29.4kb | Sense | Chr. 6 - 27936655 - 27936750 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6430711C07Rik
Genomic Map