Assay Details
Gene Symbol: 6530413G14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK018336.1 | - | 1 | 1149 | 76 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
elongation factor 1 homolog (ELF1, S. cerevisiae) |
Elof1 | 1110011K10Rik | -3.2kb | Anti-sense | Chr. 9 - 22112989 - 22114169 on NCBI Build GRCm38 |
Downstream |
acid phosphatase 5, tartrate resistant |
Acp5 |
TRACP, TRAP |
7.9kb | Anti-sense | Chr. 9 - 22126727 - 22135746 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
