Assay Details
| Gene Symbol: 8430406P12Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018390.1 | - | 1 | 853 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cyclin-dependent kinase 13 | Cdk13 |
2310015O17Rik, Cdc2l5 |
Sense | Chr. 13 - 17715962 - 17805332 on NCBI Build GRCm38 | |
| Downstream | microRNA 466i | Mir466i |
Mirn466i, mmu-mir-466i |
10.4kb | Anti-sense | Chr. 13 - 17747473 - 17747593 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

8430406P12Rik
Genomic Map