Assay Details
| Gene Symbol: 8430434A19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK018454.1 | - | 1 | 745 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC102637720 | LOC102637720 | -604.7kb | Sense | Chr. 19 - 42778653 - 42781574 on NCBI Build GRCm38 | |
| Downstream | cyclin M1 | Cnnm1 |
AI846314, Acdp1, CLP-1 |
53.0kb | Sense | Chr. 19 - 43440425 - 43497213 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

8430434A19Rik
Genomic Map