Assay Details
| Gene Symbol: Zfp952 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK050612.1 | - | 1 - 2 | 135 | 78 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 472 | Zfp472 |
KRIM-1, Krim-1A, Krim-1B, Krim1 |
-13.9kb | Sense | Chr. 17 - 32965831 - 32979211 on NCBI Build GRCm38 |
| Downstream | zinc finger protein 763 | Zfp763 |
1700065O13Rik, AI839635 |
19.3kb | Anti-sense | Chr. 17 - 33016864 - 33033381 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Zfp952
Genomic Map