Assay Details
Gene Symbol: AI314180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK054077.1 | - | 1 | 2149 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | olfactory receptor 267 | Olfr267 | MOR262-1 | -9.1kb | Sense | Chr. 4 - 58783130 - 58788108 on NCBI Build GRCm38 |
Downstream | zinc finger with KRAB and SCAN domains 16 | Zkscan16 |
XM_143800, Zfp483 |
143.9kb | Anti-sense | Chr. 4 - 58943568 - 58962624 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
