Assay Details
Gene Symbol: E230020D15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK054111.1 | - | 1 | 950 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 31463 |
Gm31463 | -52.9kb | Anti-sense | Chr. 8 - 8576522 - 8580342 on NCBI Build GRCm38 | |
Downstream | arginine and glutamate rich 1 | Arglu1 |
9430010O03Rik, C130008N12 |
31.5kb | Sense | Chr. 8 - 8666567 - 8690553 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
