Assay Details
Gene Symbol: 4930474G06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK076658.1 | - | 2 - 3 | 165 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cDNA sequence BC051408 | BC051408 | -156.1kb | Sense | Chr. 18 - 28711861 - 28726847 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA 4930578E11 gene | 4930578E11Rik | 511.5kb | Anti-sense | Chr. 18 - 29396716 - 29496416 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
