Assay Details
| Gene Symbol: Mirg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK077315.1 | - | 1 - 2 | 27 | 72 | |
| EU434920.1 | - | 1914 | 72 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 453 | Mir453 |
Mirn453, mmu-mir-453 |
-2.1kb | Sense | Chr. 12 - 109735619 - 109735700 on NCBI Build GRCm38 |
| Downstream | microRNA 541 | Mir541 |
Mirn541, mir-541, mmu-mir-541 |
906.0 | Sense | Chr. 12 - 109742409 - 109742498 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Mirg
Genomic Map