Assay Details
| Gene Symbol: Kdelc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK077468.1 | - | 1 | 1946 | 97 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | testis expressed 30 | Tex30 |
1700029F09Rik, 3110030D08Rik |
-14.3kb | Sense | Chr. 1 - 44086613 - 44102497 on NCBI Build GRCm38 |
| Downstream |
basic, immunoglobulin-like variable motif containing |
Bivm | AI854267 | 87.0 | Anti-sense | Chr. 1 - 44118888 - 44144771 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kdelc1
Genomic Map