Assay Details
| Gene Symbol: Sybu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK049811.1 | - | 2 | 3790 | 95 | |
| AK078269.1 | - | 1 | 2650 | 95 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | estrogen receptor-binding fragment-associated gene 9 | Ebag9 |
AI835379, Rcas1 |
-119.4kb | Anti-sense | Chr. 15 - 44619169 - 44641542 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 2310069G16 gene | 2310069G16Rik | 23.9kb | Anti-sense | Chr. 15 - 44787763 - 44805829 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sybu
Genomic Map