Assay Details
| Gene Symbol: 6530402D11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK078339.1 | - | 1 | 1150 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 41505 |
Gm41505 | Anti-sense | Chr. 16 - 94744212 - 94753436 on NCBI Build GRCm38 | ||
| Downstream |
predicted gene, 31641 |
Gm31641 | 433.5kb | Sense | Chr. 16 - 95184537 - 95263455 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

6530402D11Rik
Genomic Map