Assay Details
| Gene Symbol: BB131254 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079334.1 | - | 1 | 246 | 74 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cilia and flagella associated protein 74 | Cfap74 |
2010015L04Rik, A530045M11 |
Sense | Chr. 4 - 155398208 - 155466823 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 5830444B04 gene | 5830444B04Rik | 46.0 | Anti-sense | Chr. 4 - 155400969 - 155421704 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

BB131254
Genomic Map