Assay Details
| Gene Symbol: 2810454H06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_029441.1 | 1 | 489 | 64 | ||
| GenBank mRNA | AK028630.1 | - | 1 | 489 | 64 | |
| AK080385.1 | - | 1 | 56 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cAMP responsive element binding protein-like 2 | Crebl2 |
AI046348, B230205M03 |
-39.5kb | Sense | Chr. 6 - 134828686 - 134858931 on NCBI Build GRCm38 |
| Downstream | cyclin-dependent kinase inhibitor 1B | Cdkn1b |
AA408329, AI843786, Kip1, p27, p27Kip1 |
21.1kb | Sense | Chr. 6 - 134920401 - 134925525 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

2810454H06Rik
Genomic Map