Assay Details
| Gene Symbol: Sc5d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK050439.1 | - | 1 | 2195 | 69 | |
| AK080673.1 | - | 1 | 845 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA 4930546K05 gene | 4930546K05Rik | -42.4kb | Sense | Chr. 9 - 42182238 - 42209231 on NCBI Build GRCm38 | |
| Downstream | tectorin alpha | Tecta | Tctna | 76.5kb | Sense | Chr. 9 - 42329622 - 42403179 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sc5d
Genomic Map