Assay Details
Gene Symbol: 2610203C22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK082741.1 | - | 2 - 3 | 209 | 114 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 3110035E14 gene | 3110035E14Rik |
AI415019, AV164974, R75066 |
Anti-sense | Chr. 1 - 9601267 - 9627142 on NCBI Build GRCm38 | |
Downstream | myeloblastosis oncogene-like 1 | Mybl1 |
A-myb, G1-419-6, repro9 |
46.3kb | Sense | Chr. 1 - 9667407 - 9700439 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
