Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10479618 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10479622 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10479621 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10479624 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10479620 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10479619 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10479617 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10479616 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10479623 |
- |
- |
Affymetrix HT MG-430 PM Array Plate |
1438296_PM_at |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10479615 |
- |
- |
Mouse Genome 430 2.0 Array |
1438296_at |
- |
- |
Mouse Genome 430 2.0 Array |
1438296_PM_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0200023288.mm.1 |
NR_045164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023288.mm.1 |
NR_045164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023288.mm.1 |
TC0200002823.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023273.mm.1 |
NR_045164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023273.mm.1 |
NR_045164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023273.mm.1 |
TC0200002823.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023275.mm.1 |
NR_045164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023275.mm.1 |
NR_045164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023275.mm.1 |
TC0200002823.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023278.mm.1 |
NR_045164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023278.mm.1 |
NR_045164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023278.mm.1 |
TC0200002823.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023286.mm.1 |
NR_045164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023286.mm.1 |
NR_045164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023286.mm.1 |
TC0200002823.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023281.mm.1 |
NR_045164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023281.mm.1 |
NR_045164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023281.mm.1 |
TC0200002823.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023282.mm.1 |
NR_045164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023282.mm.1 |
NR_045164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023282.mm.1 |
TC0200002823.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023283.mm.1 |
NR_045164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023283.mm.1 |
NR_045164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023283.mm.1 |
TC0200002823.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023289.mm.1 |
NR_045164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023289.mm.1 |
NR_045164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023289.mm.1 |
TC0200002823.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023274.mm.1 |
NR_045164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023274.mm.1 |
NR_045164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023274.mm.1 |
TC0200002823.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023276.mm.1 |
NR_045164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023276.mm.1 |
NR_045164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023276.mm.1 |
TC0200002823.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023285.mm.1 |
NR_045164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023285.mm.1 |
NR_045164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023285.mm.1 |
TC0200002823.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023287.mm.1 |
NR_045164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023287.mm.1 |
NR_045164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023287.mm.1 |
TC0200002823.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023279.mm.1 |
NR_045164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023279.mm.1 |
NR_045164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023279.mm.1 |
TC0200002823.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023280.mm.1 |
NR_045164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023280.mm.1 |
NR_045164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023280.mm.1 |
TC0200002823.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023284.mm.1 |
NR_045164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023284.mm.1 |
NR_045164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023284.mm.1 |
TC0200002823.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200023277.mm.1 |
NR_045164.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023277.mm.1 |
NR_045164.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200023277.mm.1 |
TC0200002823.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1438296_at |
- |
- |
Mouse Expression Set 430 |
1438296_PM_at |
- |
- |
Murine Genome U74v2 Set |
167982_i_at |
- |
- |
Murine Genome U74v2 Set |
167982_PM_i_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1438296_at |
- |
- |
Affymetrix HT Mouse Genome 430B Array |
1438296_PM_at |
- |
- |