Assay Details
Gene Symbol: D130017N08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK083834.1 | - | 2978 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Downstream | eukaryotic translation initiation factor 2 alpha kinase 1 | Eif2ak1 |
HCR, Hri |
105.2kb | Sense | Chr. 5 - 143871862 - 143902717 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
