Assay Details
| Gene Symbol: Usp29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK086482.1 | - | 1 | 2151 | 90 | |
| BC082334.1 | - | 3 | 3595 | 90 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | paternally expressed 3 | Peg3 |
AL022617, ASF-1, End4, Gcap4, Pw1, Zfp102, mKIAA0287 |
-36.8kb | Anti-sense | Chr. 7 - 6705960 - 6730419 on NCBI Build GRCm38 |
| Downstream | microRNA 3099 | Mir3099 |
mir-3099, mmu-mir-3099 |
33.5kb | Sense | Chr. 7 - 6803587 - 6803670 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Usp29
Genomic Map