Assay Details
| Gene Symbol: A730091E23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK043390.1 | - | 1 | 660 | 97 | |
| AK086836.1 | - | 1 | 649 | 97 | ||
| BB529729.2 | - | 1 | 293 | 97 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
GDP-mannose 4, 6-dehydratase |
Gmds |
BC031788, C87208 |
-106.0 | Anti-sense | Chr. 13 - 31819583 - 32338725 on NCBI Build GRCm38 |
| Downstream |
myosin light chain kinase family, member 4 |
Mylk4 | EG238564 | 361.0kb | Anti-sense | Chr. 13 - 32700827 - 32806771 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

A730091E23Rik
Genomic Map