Assay Details
| Gene Symbol: Nsun3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK088718.1 | - | 1 | 1455 | 65 | |
| AK162988.1 | - | 1 | 1446 | 65 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Eph receptor A6 | Epha6 |
Ehk2, Hek12, m-ehk2 |
-2.2mb | Sense | Chr. 16 - 59653061 - 60606484 on NCBI Build GRCm38 |
| Downstream | ADP-ribosylation factor-like 13B | Arl13b |
A530097K21Rik, A930014M17Rik, Arl2l1, C530009C10Rik, hnn |
6.9kb | Sense | Chr. 16 - 62793678 - 62847055 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nsun3
Genomic Map