Assay Details
| Gene Symbol: Rnf165 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164504.1 | NP_001157976.1 | 8 | 5456 | 68 | |
| GenBank mRNA | AK090360.1 | - | 1 | 1047 | 68 | |
| BC066016.1 | - | 1 | 1246 | 68 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | lipoxygenase homology domains 1 | Loxhd1 |
1700096C21Rik, 4932417K07, sba |
-14.9kb | Anti-sense | Chr. 18 - 77281949 - 77442932 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 8030462N17 gene | 8030462N17Rik |
Akd2, Ark2N |
172.5kb | Sense | Chr. 18 - 77631488 - 77714489 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Rnf165
Genomic Map