Assay Details
Gene Symbol: 4930578E11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045391.1 | 7 - 8 | 1665 | 78 | ||
GenBank mRNA | AK132929.1 | - | 7 - 8 | 1666 | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4930474G06 gene | 4930474G06Rik | -398.5kb | Anti-sense | Chr. 18 - 28560164 - 28998176 on NCBI Build GRCm38 | |
Downstream | RIKEN cDNA 4930527G23 gene | 4930527G23Rik | 2.0kb | Anti-sense | Chr. 18 - 29442200 - 29466107 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2110532 | AK132929.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 225522 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 12247 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 744067 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 790479 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 298653 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 122519 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 875989 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 746624 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 584551 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 143976 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 250374 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 723951 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 14976 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 743142 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 940163 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 63674 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 219359 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352609 | AK151902.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352609 | BC131964.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352609 | BC131966.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 352609 | NM_148939.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 904601 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 721644 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 822997 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 298382 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 757885 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 852841 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 432511 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 843444 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 72769 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669390 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 266530 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 48360 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 76825 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 826103 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 310990 | AK038283.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107498 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 678360 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 214224 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 90754 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35218 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 26251 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 18986 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110532 | AK132929.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08950256 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08950174 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08950327 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08950199 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08950311 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08950297 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03439432 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03439381 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08950358 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08950210 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08950237 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P110382 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03439432 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08950297 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08950199 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08950237 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13776280 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13776282 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587597 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587689 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587701 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587643 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587637 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587634 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587714 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587708 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587178 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587626 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587734 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587720 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587677 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587737 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587297 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587608 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587681 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587738 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587731 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14098058 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587647 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13917027 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587143 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587598 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587179 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587594 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587254 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587269 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14135257 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587151 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587243 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587727 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587710 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587264 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587603 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13998331 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587685 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587680 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587182 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587592 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587300 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587696 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587702 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587145 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13917019 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587725 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587728 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587674 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587157 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587706 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14098048 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587177 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587292 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587739 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13997645 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587148 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587676 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587613 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587268 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587339 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587293 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587646 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587291 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587611 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587590 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587240 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587705 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587723 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13859055 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14098052 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587295 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587711 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587247 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587707 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587703 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587332 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587688 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587188 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587257 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587686 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587272 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587256 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587741 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587602 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13917024 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587271 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587245 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587259 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587335 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13687067 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13862293 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587640 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587628 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587745 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587195 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587736 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587181 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587713 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587636 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14098062 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587252 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587675 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587691 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587724 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13889487 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587301 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587683 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587193 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587588 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587632 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587644 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13639915 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587187 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587692 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587642 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587277 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587294 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587328 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587330 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587694 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587270 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587273 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587155 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13714631 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587146 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587630 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587648 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587684 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587275 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587249 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587331 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587340 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587718 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587758 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587154 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587610 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587267 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587260 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587326 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587650 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587699 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14098050 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587744 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587158 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587333 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587144 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587190 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587304 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587184 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587296 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587678 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31587276 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR1800008042.mm.1 | NR_045391.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800008042.mm.1 | NR_045391.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800008042.mm.1 | TC1800001101.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800008041.mm.1 | NR_045391.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800008041.mm.1 | NR_045391.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800008041.mm.1 | TC1800001101.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800008043.mm.1 | NR_045391.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800008043.mm.1 | NR_045391.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800008043.mm.1 | TC1800001101.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 105890601 | - | - |
