Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00019187.mm.1 |
NR_045455.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019187.mm.1 |
NR_045455.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019187.mm.1 |
NR_045456.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019187.mm.1 |
NR_045456.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019187.mm.1 |
TC0X00003059.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00019189.mm.1 |
NR_045455.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019189.mm.1 |
NR_045455.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00019189.mm.1 |
TC0X00003059.mm.1 |
MTA TC |