Assay Details
| Gene Symbol: E030022I16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK134478.1 | - | 1 | 2250 | 100 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DEAD (Asp-Glu-Ala-Asp) box polypeptide 6 | Ddx6 |
1110001P04Rik, E230023J21Rik, HLR2, mRCK/P54, p54, rck |
Sense | Chr. 9 - 44602716 - 44640731 on NCBI Build GRCm38 | |
| Downstream | trehalase (brush-border membrane glycoprotein) | Treh | 2210412M19Rik | 52.2kb | Sense | Chr. 9 - 44673233 - 44686305 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

E030022I16Rik
Genomic Map