Assay Details
Gene Symbol: AI256396 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK046686.1 | - | 2522 | 96 | ||
AK134520.1 | - | 1457 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | glucoside xylosyltransferase 2 | Gxylt2 |
Glt8d4, Gm152 |
-3.9kb | Sense | Chr. 6 - 100704293 - 100805686 on NCBI Build GRCm38 |
Downstream |
protein phosphatase 4, regulatory subunit 2 |
Ppp4r2 |
BE691708, C230060M08Rik |
23.5kb | Sense | Chr. 6 - 100833623 - 100869934 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
