Assay Details
| Gene Symbol: Gm7759 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK135850.1 | - | 2 - 3 | 217 | 94 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | regulating synaptic membrane exocytosis 1 | Rims1 |
C030033M19Rik, RIM1, RIM1a, RIM1alpha, Rab3ip1, Rim, Serg1, mKIAA0340 |
-97.7kb | Anti-sense | Chr. 1 - 22468191 - 22805724 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA 4933415F23 gene | 4933415F23Rik | 195.4kb | Anti-sense | Chr. 1 - 23100473 - 23102253 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm7759
Genomic Map