Assay Details
Gene Symbol: Gm7056 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK136178.1 | - | 1 - 2 | 137 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | neuropeptide FF receptor 2 | Npffr2 |
Gpr74, HG31, NPFF2 |
-32.5kb | Sense | Chr. 5 - 89526577 - 89583914 on NCBI Build GRCm38 |
Downstream |
a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 3 |
Adamts3 |
1100001H14Rik, 6330442E02Rik |
53.8kb | Anti-sense | Chr. 5 - 89673841 - 89883669 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
