Assay Details
Gene Symbol: Gm816 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK136795.1 | - | 1 - 2 | 253 | 120 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 6343 | Mir6343 |
Gm27304, mmu-mir-6343 |
-380.8kb | Anti-sense | Chr. 1 - 76509560 - 76509643 on NCBI Build GRCm38 |
Downstream | Eph receptor A4 | Epha4 |
2900005C20Rik, AI385584, Cek8, Hek8, Sek, Sek1, Tyro1, rb |
339.4kb | Sense | Chr. 1 - 77367182 - 77515103 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 971283 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 877657 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 970093 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267674 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 261899 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 234154 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526488 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 226484 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 939026 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 603258 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 720491 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 145403 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 591521 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 766783 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 832882 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 567338 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947331 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 55500 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335899 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 184316 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750513 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 847354 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 928885 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 402786 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441263 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 385025 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 44923 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 406376 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 762873 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 285157 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 482742 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 15531 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 282076 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 71628 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 890028 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 390404 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 934491 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 507546 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 697877 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 747990 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 218992 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 472781 | AK002549.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 472781 | BC019786.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 472781 | BC037080.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 472781 | BC089608.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 472781 | NM_007860.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 472781 | U49861.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 205231 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206314 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 459857 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537824 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 674472 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 909048 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 17465 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 116928 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 3708 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 73869 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 11966 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 135519 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 73074 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35929 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 26101 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 130033 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 168832 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 114958 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04154120 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04153934 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00108640 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04153886 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04153990 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04154148 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00108539 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04154188 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04154036 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04153990 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04153934 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04154120 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P04154036 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20363083 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362810 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362380 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362813 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00646336 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00965024 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362187 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362723 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362809 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362104 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362391 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20363090 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362107 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362326 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362756 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362656 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362715 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362200 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00208743 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362377 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362353 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362335 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20363114 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362100 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362805 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20363072 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362385 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362815 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362873 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01095778 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362336 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362822 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362659 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00384033 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00797169 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362883 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00024860 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362654 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20363087 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362387 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00440433 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362876 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362349 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362379 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362879 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362206 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00672730 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362389 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362637 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00428435 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362814 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362657 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362203 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362204 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00080438 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362652 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362882 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362658 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00440432 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362634 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362870 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362720 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362820 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362647 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00797167 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362199 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01077053 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362344 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362754 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362703 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362113 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362197 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362384 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362382 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00767438 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00440429 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362806 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362098 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362105 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362342 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20363088 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00769673 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00545902 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362724 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362826 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362740 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362635 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362708 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362182 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362397 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01135684 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362339 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362646 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362744 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362875 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362880 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362334 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20363089 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362752 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01077059 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01142993 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362758 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362180 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362319 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00745202 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362214 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362871 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362378 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362709 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362177 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362653 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362354 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362375 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20363117 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20363070 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362400 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362208 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362331 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362719 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362741 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362186 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362330 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362207 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01142992 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362639 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362751 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362743 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362317 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362399 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362889 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362759 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362748 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20363076 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362322 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362721 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362648 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362394 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362712 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362811 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362194 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362701 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362760 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00829367 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00320388 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362633 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362641 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362396 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362321 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362185 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362183 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362350 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00829371 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20363080 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362327 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362346 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362373 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20363085 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362101 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362191 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362704 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362705 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00452470 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00452457 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362401 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20363116 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362807 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362717 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00166924 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20362210 | - | - |
